Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021658.1 | NP_067690.1 | 5, 6 | 1972 | ||
GenBank mRNA | AF247453.1 | - | 1972 |