Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022236.1 | NP_071572.1 | 2 | 372 | ||
GenBank mRNA | AB027155.1 | - | 2 | 372 | ||
AB027156.1 | - | 707 | ||||
AY462091.1 | - | 338 | ||||
AY462092.1 | - | 446 | ||||
AY462093.1 | - | 484 | ||||
AY462095.1 | - | 619 |