Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022385.3 | NP_071780.1 | 3, 4 | 325 | ||
GenBank mRNA | BC061553.1 | - | 408 | 6889396 | ||
FQ226660.1 | - | 3, 4 | 381 | |||
U12402.1 | - | 3, 4 | 257 | |||
X76920.1 | - | 3, 4 | 325 |