Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022798.2 | NP_073635.1 | 3 | 506 | ||
GenBank mRNA | AF156272.1 | - | 3 | 613 | ||
BC078819.1 | - | 3 | 506 | 7113511 |