Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024361.1 | NP_077337.1 | 2, 3 | 967 | ||
GenBank mRNA | M92042.1 | - | 2, 3 | 967 |