Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celsr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031320.1 | NP_112610.1 | 1 | 530 | ||
XM_006243860.2 | XP_006243922.1 | 1 | 267 | |||
XM_006243861.2 | XP_006243923.1 | 1 | 267 | |||
GenBank mRNA | AB011528.1 | - | 1 | 530 |
Set Membership: |
G-protein coupled receptor |