Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UBA52 SIRNA Assays ›
Gene Symbol:
Uba52
Gene Name:
ubiquitin A-52 residue ribosomal protein fusion product 1
Gene Aliases:
Rps27a, Ubb
Chromosome Location:
Chr.16: 20668941 - 20671127 on Build Rnor_6.0
UniGene ID:
Rn.110618
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
53926
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Uba52
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031687.3
NP_113875.1
2
138
XM_006252934.2
XP_006252996.1
2
225
XM_006252935.1
XP_006252997.1
2
343
XM_008771145.1
XP_008769367.1
2
221
XM_008771146.1
XP_008769368.1
2
165
GenBank mRNA
BC061544.1
-
2
134
6922034
BP492585.1
-
1
24
FQ209944.1
-
2
130
FQ210541.1
-
2
127
FQ210904.1
-
2
128
FQ212001.1
-
2
132
FQ212205.1
-
2
130
FQ212291.1
-
2
128
FQ212810.1
-
2
131
FQ217440.1
-
2
123
FQ218121.1
-
2
124
FQ219376.1
-
2
129
FQ219803.1
-
2
130
FQ221045.1
-
2
127
FQ221118.1
-
2
128
FQ221238.1
-
2
348
FQ221621.1
-
2
124
FQ221631.1
-
2
130
FQ221755.1
-
2
128
FQ221772.1
-
2
128
FQ221777.1
-
2
128
FQ222045.1
-
2
128
FQ222098.1
-
2
124
FQ222371.1
-
2
127
FQ222440.1
-
2
139
FQ222483.1
-
2
127
FQ222507.1
-
2
122
FQ222550.1
-
2
124
FQ222700.1
-
2
127
FQ223258.1
-
2
130
FQ223423.1
-
2
124
FQ223456.1
-
2
125
FQ224647.1
-
1
118
FQ224702.1
-
2
128
FQ228362.1
-
2
124
FQ228566.1
-
2
128
X82636.1
-
1
100
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019883, EF437347, AK018726, BY726472, AF118402, and others.
Gene Ontology Categories:
Function(s)
Process(es)
translation
structural constituent of ribosome
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0