Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrsp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031714.1 | NP_113902.1 | 3 | 273 | ||
GenBank mRNA | AF015949.1 | - | 3 | 273 | ||
BC078779.1 | - | 3 | 264 | 7128728 | ||
FQ209529.1 | - | 3 | 286 | |||
FQ209945.1 | - | 2 | 123 | |||
FQ210021.1 | - | 3 | 278 | |||
FQ218425.1 | - | 3 | 280 | |||
FQ218811.1 | - | 3 | 281 | |||
FQ219440.1 | - | 3 | 280 | |||
FQ219496.1 | - | 3 | 286 | |||
X70825.1 | - | 3 | 241 |