Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053437.1 | NP_445889.1 | 11, 12 | 1012 | ||
GenBank mRNA | AB062759.1 | - | 11, 12 | 990 | ||
AB062760.1 | - | 11, 12 | 990 | |||
AB062761.1 | - | 11, 12 | 990 | |||
AB062762.1 | - | 11, 12 | 990 | |||
AB062763.1 | - | 11, 12 | 988 | |||
AF296131.1 | - | 11, 12 | 1012 | |||
AJ345014.1 | - | 5, 6 | 300 | |||
BC081742.1 | - | 11, 12 | 984 |