Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199492.1 | NP_001186421.1 | 3 | 426 | ||
NM_014456.4 | NP_055271.2 | 3 | 468 | |||
NM_145341.3 | NP_663314.1 | 4 | 552 | |||
GenBank mRNA | AK223623.1 | - | 3 | 342 | ||
AK296753.1 | - | 3 | 293 | |||
AK312540.1 | - | 3 | 364 | |||
AK315295.1 | - | 4 | 430 | |||
BC015036.2 | - | 3 | 332 | |||
BC026104.2 | - | 3 | 335 | 4798293 | ||
BC031049.1 | - | 3 | 335 | 5277621 | ||
BI910668.1 | - | 3 | 430 | |||
BX537500.1 | - | 2 | 3416 | |||
BX647834.1 | - | 3 | 312 | |||
DB241608.1 | - | 3 | 469 | |||
U96628.1 | - | 3 | 266 |