Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oplah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053904.1 | NP_446356.1 | 22 | 3170 | ||
XM_006241771.2 | XP_006241833.1 | 22 | 4830 | |||
XM_006241772.2 | XP_006241834.1 | 22 | 4827 | |||
XM_006241773.2 | XP_006241835.1 | 22 | 4713 | |||
XM_006241774.2 | XP_006241836.1 | 22 | 3508 | |||
XM_006241775.2 | XP_006241837.1 | 23 | 3367 | |||
GenBank mRNA | BC085330.1 | - | 22 | 3235 | 7096390 | |
U70825.1 | - | 22 | 3170 |