Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057188.1 | NP_476536.1 | 4 | 451 | ||
GenBank mRNA | AF090867.1 | - | 4 | 451 |