Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130430.1 | NP_569114.1 | 1 | 604 | ||
GenBank mRNA | AF067728.1 | - | 1 | 604 | ||
FQ214178.1 | - | 148 | ||||
FQ225583.1 | - | 137 | ||||
FQ227319.1 | - | 137 | ||||
FQ229304.1 | - | 170 |