Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BFAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016561.2 | NP_057645.1 | 2, 3 | 552 | ||
GenBank mRNA | AF173003.1 | - | 2, 3 | 424 | ||
AK291114.1 | - | 2, 3 | 360 | |||
AK297067.1 | - | 2, 3 | 360 | |||
AK313890.1 | - | 1, 2 | 360 | |||
BC003054.1 | - | 2, 3 | 383 | 3537783 | ||
BI912683.1 | - | 2, 3 | 552 |