Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Daxx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080891.2 | NP_543167.2 | 5, 6 | 1536 | ||
XM_006256103.2 | XP_006256165.1 | 5, 6 | 2318 | |||
XM_006256104.2 | XP_006256166.1 | 5, 6 | 1992 | |||
XM_006256105.2 | XP_006256167.1 | 5, 6 | 1888 | |||
XM_006256106.2 | XP_006256168.1 | 5, 6 | 1554 | |||
XM_006256107.2 | XP_006256169.1 | 5, 6 | 1658 | |||
XM_006256108.2 | XP_006256170.1 | 5, 6 | 1538 | |||
XM_006256109.1 | XP_006256171.1 | 4, 5 | 2411 | |||
XM_006256110.2 | XP_006256172.1 | 5, 6 | 1537 | |||
XM_008772704.1 | XP_008770926.1 | 4, 5 | 2411 | |||
GenBank mRNA | AB064671.1 | - | 4, 5 | 1440 | ||
BC128729.1 | - | 5, 6 | 1536 | 8366304 | ||
FQ223571.1 | - | 5, 6 | 1507 |