Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134405.1 | NP_599232.1 | 12 | 2019 | ||
GenBank mRNA | AB023645.1 | - | 12 | 2019 | ||
BC081728.1 | - | 13 | 2052 | 7129648 |