Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnrc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138845.2 | NP_620200.2 | 25 | 5438 | ||
XM_006242061.2 | XP_006242123.1 | 22 | 5718 | |||
XM_006242063.2 | XP_006242125.1 | 21 | 5546 | |||
XM_006242064.2 | XP_006242126.1 | 20 | 5387 | |||
GenBank mRNA | AF275151.1 | - | 17 | 2193 | ||
BC061751.1 | - | 9 | 1114 | |||
FQ226824.1 | - | 2 | 308 |