Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APOE SIRNA Assays ›
Gene Symbol:
Apoe
Gene Name:
apolipoprotein E
Gene Aliases:
APOEA
Chromosome Location:
Chr.1: 80612894 - 80616820 on Build Rnor_6.0
UniGene ID:
Rn.32351
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
57850
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Apoe
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270681.1
NP_001257610.1
4
390
NM_001270682.1
NP_001257611.1
4
369
NM_001270683.1
NP_001257612.1
4
356
NM_001270684.1
NP_001257613.1
4
332
NM_138828.3
NP_620183.2
4
350
GenBank mRNA
BC060313.1
-
4
323
6919048
BC086581.1
-
4
293
7307758
FQ209421.1
-
4
315
FQ209426.1
-
4
321
FQ209428.1
-
4
319
FQ209457.1
-
4
318
FQ209625.1
-
4
319
FQ209879.1
-
3
266
FQ209926.1
-
4
319
FQ210262.1
-
4
319
FQ210410.1
-
4
319
FQ211250.1
-
4
320
FQ212760.1
-
4
319
FQ212769.1
-
4
319
FQ213060.1
-
4
315
FQ213396.1
-
4
315
FQ213533.1
-
4
322
FQ213898.1
-
4
311
FQ214072.1
-
4
319
FQ214282.1
-
4
319
FQ218386.1
-
4
318
FQ218473.1
-
4
348
FQ218485.1
-
4
319
FQ218699.1
-
4
321
FQ218898.1
-
4
321
FQ218953.1
-
3
375
FQ219009.1
-
4
319
FQ219124.1
-
3
279
FQ219144.1
-
4
319
FQ219202.1
-
3
270
FQ219319.1
-
4
319
FQ219432.1
-
4
319
FQ219448.1
-
4
321
FQ219475.1
-
4
314
FQ219490.1
-
4
319
FQ219516.1
-
4
319
FQ219534.1
-
4
319
FQ219700.1
-
4
319
FQ219711.1
-
4
319
FQ220125.1
-
4
315
FQ220683.1
-
4
319
FQ220752.1
-
4
350
FQ220879.1
-
4
323
FQ221856.1
-
4
319
FQ223299.1
-
3
270
FQ229186.1
-
4
319
FQ229201.1
-
4
319
FQ229530.1
-
3
270
FQ229565.1
-
4
315
FQ229650.1
-
4
319
FQ229759.1
-
4
319
FQ229950.1
-
4
319
FQ230084.1
-
4
319
FQ231375.1
-
4
348
J00705.1
-
3
277
Back To Top
More Information
Panther Classification:
Molecular Function -
apolipoprotein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of endothelial cell proliferation
response to dietary excess
lipid metabolic process
triglyceride metabolic process
cholesterol biosynthetic process
cholesterol catabolic process
lipid transport
cellular calcium ion homeostasis
receptor-mediated endocytosis
response to oxidative stress
G-protein coupled receptor signaling pathway
nitric oxide mediated signal transduction
aging
protein localization
cholesterol metabolic process
regulation of gene expression
negative regulation of platelet activation
positive regulation of cholesterol esterification
positive regulation of cholesterol efflux
peripheral nervous system axon regeneration
long-chain fatty acid transport
protein import
triglyceride catabolic process
cGMP-mediated signaling
negative regulation of blood coagulation
positive regulation of cGMP biosynthetic process
neuron projection regeneration
axon regeneration
regulation of cholesterol transport
regulation of Cdc42 protein signal transduction
response to retinoic acid
positive regulation of low-density lipoprotein particle receptor catabolic process
response to insulin
cholesterol efflux
phospholipid efflux
very-low-density lipoprotein particle remodeling
low-density lipoprotein particle remodeling
high-density lipoprotein particle remodeling
high-density lipoprotein particle assembly
chylomicron remnant clearance
high-density lipoprotein particle clearance
very-low-density lipoprotein particle clearance
lipoprotein metabolic process
lipoprotein biosynthetic process
lipoprotein catabolic process
vasodilation
cholesterol homeostasis
positive regulation of catalytic activity
negative regulation of MAP kinase activity
negative regulation of neuron apoptotic process
negative regulation of blood vessel endothelial cell migration
reverse cholesterol transport
response to ethanol
negative regulation of cholesterol biosynthetic process
positive regulation of axon extension
positive regulation of lipid biosynthetic process
oligodendrocyte differentiation
artery morphogenesis
negative regulation of inflammatory response
positive regulation of nitric-oxide synthase activity
positive regulation of membrane protein ectodomain proteolysis
negative regulation of lipid biosynthetic process
maintenance of location in cell
lipid homeostasis
fatty acid homeostasis
positive regulation of dendritic spine development
negative regulation of dendritic spine development
cellular response to interleukin-1
cellular response to growth factor stimulus
cellular response to cholesterol
cardiovascular system development
negative regulation of cholesterol efflux
AMPA glutamate receptor clustering
NMDA glutamate receptor clustering
regulation of beta-amyloid clearance
regulation of neuron death
negative regulation of neuron death
positive regulation of neuron death
negative regulation of postsynaptic membrane organization
positive regulation of postsynaptic membrane organization
negative regulation of presynaptic membrane organization
positive regulation of presynaptic membrane organization
positive regulation of beta-amyloid formation
negative regulation of beta-amyloid formation
regulation of tau-protein kinase activity
negative regulation of dendritic spine maintenance
positive regulation of dendritic spine maintenance
positive regulation of phospholipid efflux
positive regulation of neurofibrillary tangle assembly
negative regulation of phospholipid efflux
negative regulation of lipid transport across blood brain barrier
positive regulation of lipid transport across blood brain barrier
beta-amyloid binding
receptor binding
lipid transporter activity
phospholipid binding
heparin binding
lipid binding
cholesterol binding
antioxidant activity
cholesterol transporter activity
identical protein binding
protein homodimerization activity
hydroxyapatite binding
metal chelating activity
tau protein binding
low-density lipoprotein particle receptor binding
phosphatidylcholine-sterol O-acyltransferase activator activity
very-low-density lipoprotein particle receptor binding
lipoprotein particle binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0