Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145776.1 | NP_665719.1 | 13 | 1179 | ||
XM_006243708.2 | XP_006243770.1 | 13 | 1158 | |||
GenBank mRNA | AF273025.1 | - | 13 | 1179 | ||
BC081717.1 | - | 13 | 1200 | 7127614 |