Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007481.3 | NP_031507.1 | 2 | 1108 | ||
GenBank mRNA | AK006366.1 | - | 1 | 818 | ||
AK156960.1 | - | 2 | 1093 | |||
BC003478.1 | - | 2 | 1070 | 2655889 | ||
BC083112.1 | - | 2 | 1108 | 5354797 | ||
D87903.1 | - | 2 | 1050 |