Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007683.3 | NP_031709.2 | 3, 4 | 384 | ||
GenBank mRNA | AK011004.1 | - | 3, 4 | 353 | ||
AK137803.1 | - | 3, 4 | 353 | |||
AK137953.1 | - | 3, 4 | 350 | |||
AK143566.1 | - | 3, 4 | 385 | |||
BC150700.1 | - | 3, 4 | 180 | 9087611 | ||
U03113.1 | - | 3, 4 | 270 |