Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crygc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082573.2 | NP_001076042.1 | 3 | 336 | ||
NM_007775.2 | NP_031801.1 | 3 | 333 | |||
GenBank mRNA | BC056454.1 | - | 2 | 312 | 6824225 | |
CJ066821.1 | - | 3 | 334 | |||
EF426301.1 | - | 3 | 322 | |||
EF426302.1 | - | 3 | 325 | |||
M64544.1 | - | 2 | 285 |