Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB560752.1 | - | 2, 3 | 680 | ||
AB560753.1 | - | 1, 2 | 543 | |||
AB560754.1 | - | 1, 2 | 543 | |||
AB560755.1 | - | 1, 2 | 543 | |||
AB560756.1 | - | 1, 2 | 543 | |||
AK036627.1 | - | 2, 3 | 654 | |||
AK039911.1 | - | 5, 6 | 1157 | |||
AK041525.1 | - | 3, 4 | 802 | |||
AK077236.1 | - | 2, 3 | 659 | |||
AK087638.1 | - | 2, 3 | 632 | |||
AK139745.1 | - | 3, 4 | 1214 | |||
EU791538.1 | - | 2, 3 | 598 | |||
EU791540.1 | - | 2, 3 | 723 | |||
M38651.1 | - | 2, 3 | 662 |