Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007995.3 | NP_032021.1 | 10 | 1214 | ||
GenBank mRNA | AB007813.1 | - | 10 | 1106 | ||
AK133539.1 | - | 10 | 1191 | |||
AK138752.1 | - | 10 | 1161 | |||
AK159651.1 | - | 10 | 1184 | |||
BC019180.1 | - | 10 | 1162 | 5008934 |