Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293623.1 | NP_001280552.1 | 5 | 1217 | ||
NM_001293624.1 | NP_001280553.1 | 5 | 988 | |||
NM_001293625.1 | NP_001280554.1 | 5 | 984 | |||
NM_008253.4 | NP_032279.1 | 5 | 1170 | |||
GenBank mRNA | AF022465.1 | - | 4 | 918 | ||
AK032163.1 | - | 5 | 1094 | |||
AK141399.1 | - | 5 | 987 | |||
AK146795.1 | - | 5 | 987 | |||
AK159119.1 | - | 5 | 961 | |||
AK159322.1 | - | 5 | 961 | |||
BC011276.1 | - | 5 | 951 | 3980194 | ||
BC083352.1 | - | 4 | 902 | 30540227 | ||
BC158037.1 | - | 5 | 922 | 9007340 | ||
Y10044.1 | - | 1 | 310 |