Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195046.1 | NP_001181975.1 | 5, 6 | 570 | ||
NM_001195047.1 | NP_001181976.1 | 4, 5 | 525 | |||
NM_001195048.1 | NP_001181977.1 | 5, 6 | 680 | |||
NM_001195049.1 | NP_001181978.1 | 7, 8 | 944 | |||
NM_008778.3 | NP_032804.2 | 4, 5 | 553 | |||
GenBank mRNA | AF082297.1 | - | 3, 4 | 467 | ||
AJ496262.1 | - | 3, 4 | 435 | |||
AJ496263.1 | - | 4, 5 | 480 | |||
AK031853.1 | - | 5, 6 | 561 | |||
AM943853.1 | - | 4, 5 | 498 | |||
AM943854.1 | - | 5, 6 | 543 | |||
BC053403.1 | - | 4, 5 | 560 | 30060082 | ||
U39738.1 | - | 7, 8 | 951 |