Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008859.2 | NP_032885.1 | 3 | 299 | ||
GenBank mRNA | AK037664.1 | - | 3 | 371 | ||
AK085546.1 | - | 3 | 569 | |||
BC138552.1 | - | 2 | 230 | 8861572 | ||
BC145769.1 | - | 2 | 230 | 40131072 | ||
D11091.1 | - | 3 | 299 |