Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009063.3 | NP_033089.2 | 4 | 349 | ||
GenBank mRNA | AK004165.1 | - | 4 | 350 | ||
AK028765.1 | - | 351 | ||||
AK030300.1 | - | 351 | ||||
AK044096.1 | - | 4 | 350 | |||
AK054098.1 | - | 4 | 350 | |||
BC037683.1 | - | 4 | 336 | 3962083 | ||
U67188.1 | - | 306 |