Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selplg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009151.3 | NP_033177.3 | 2 | 829 | ||
GenBank mRNA | AK089188.1 | - | 2 | 805 | ||
AK089214.1 | - | 2 | 755 | |||
AK155762.1 | - | 2 | 762 | |||
AK156933.1 | - | 2 | 760 | |||
AK158134.1 | - | 2 | 791 | |||
AK171693.1 | - | 2, 3 | 593 | |||
AK172077.1 | - | 2 | 763 | |||
BC003874.1 | - | 3 | 701 | 3495181 | ||
BE859840.1 | - | 1 | 329 | |||
BG083864.2 | - | 1 | 30 | |||
X91144.1 | - | 2 | 618 |