Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082552.2 | NP_001076021.1 | 2 | 317 | ||
NM_009277.4 | NP_033303.3 | 3 | 386 | |||
GenBank mRNA | AK141294.1 | - | 3 | 365 | ||
AK150484.1 | - | 2 | 281 | |||
AK152619.1 | - | 2 | 281 | |||
AK171489.1 | - | 3 | 350 | |||
BB661457.1 | - | 3 | 373 | |||
BC010580.1 | - | 3 | 326 | 3584654 | ||
CJ051157.1 | - | 3 | 388 | |||
CT010336.1 | - | 1 | 211 | |||
L27990.1 | - | 3 | 351 |