Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166480.1 | NP_001159952.1 | 3 | 463 | ||
NM_009279.3 | NP_033305.1 | 3 | 345 | |||
GenBank mRNA | AK003044.1 | - | 3 | 325 | ||
AK007921.1 | - | 3 | 301 | |||
AK013723.1 | - | 3 | 307 | |||
AK142600.1 | - | 1 | 2745 | |||
AK160089.1 | - | 3 | 311 | |||
AK191259.1 | - | 1 | 45 | |||
BC008172.1 | - | 3 | 266 | 3481948 | ||
BC012971.1 | - | 2 | 1846 | |||
BC019419.1 | - | 2 | 1796 | |||
BI989805.1 | - | 3 | 463 | |||
BQ747160.1 | - | 3 | 248 | |||
CJ144362.1 | - | 3 | 350 | |||
X90582.1 | - | 3 | 426 |