Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016541.2 | NP_057625.1 | 3 | 293 | ||
GenBank mRNA | AB030207.1 | - | 3 | 272 | ||
AF493880.1 | - | 2 | 191 | |||
AK312139.1 | - | 3 | 294 | |||
AY029486.1 | - | 3 | 259 | |||
BC093760.1 | - | 2 | 224 | 7939605 | ||
BC095525.1 | - | 2 | 194 | 6519181 | ||
DA111448.1 | - | 3 | 294 |