Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009274.2 | NP_033300.2 | 5, 6 | 675 | ||
GenBank mRNA | AB006036.1 | - | 5, 6 | 468 | ||
AK136861.1 | - | 5, 6 | 641 | |||
AK161384.1 | - | 6, 7 | 734 | |||
AK169580.1 | - | 5, 6 | 648 | |||
AK178771.1 | - | 154 | ||||
BC020178.1 | - | 5, 6 | 572 | 4507346 | ||
BC062941.1 | - | 6, 7 | 620 | |||
U92456.1 | - | 6, 7 | 597 |