Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009536.4 | NP_033562.3 | 2 | 356 | ||
GenBank mRNA | AF483478.1 | - | 2 | 215 | ||
AF483479.1 | - | 2 | 215 | |||
AK075995.1 | - | 2 | 305 | |||
AK135831.1 | - | 2 | 284 | |||
AK135867.1 | - | 2 | 284 | |||
AK169812.1 | - | 2 | 311 | |||
AK192964.1 | - | 1 | 114 | |||
AK202856.1 | - | 1 | 114 | |||
AK212566.1 | - | 1 | 114 | |||
AK214161.1 | - | 1 | 114 | |||
AK218792.1 | - | 1 | 114 | |||
AK220088.1 | - | 1 | 114 | |||
AX775991.1 | - | 2 | 359 | |||
AX775993.1 | - | 1 | 152 | |||
BC058686.1 | - | 2 | 315 | 6411635 | ||
D87663.1 | - | 2 | 243 | |||
Z19599.1 | - | 2 | 298 |