Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009551.5 | NP_033577.1 | 6 | 1291 | ||
GenBank mRNA | AF062071.1 | - | 5 | 883 | ||
AK076397.1 | - | 6 | 1292 | |||
AK131911.1 | - | 6 | 1287 | |||
AK185148.1 | - | 1 | 301 | |||
BC061693.1 | - | 2 | 4035 | |||
BC080270.1 | - | 6 | 1272 | |||
BC107566.1 | - | 6 | 855 | 5389770 |