Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lefty1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010094.3 | NP_034224.1 | 1 | 214 | ||
GenBank mRNA | AJ000082.1 | - | 1 | 194 | ||
AK131943.1 | - | 1 | 216 | |||
BC050221.1 | - | 1 | 177 | 30005856 | ||
D83921.1 | - | 1 | 231 | |||
Z73151.1 | - | 1 | 194 |