Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010127.3 | NP_034257.1 | 5, 6 | 1282 | ||
GenBank mRNA | AB221558.1 | - | 4, 5 | 572 | ||
AK006732.1 | - | 1, 2 | 362 | |||
AY259046.1 | - | 9, 10 | 2416 | |||
AY259047.1 | - | 8, 9 | 2162 | |||
BC053046.1 | - | 7, 8 | 1112 | |||
BC059830.1 | - | 5, 6 | 1282 | 6410550 | ||
BC085139.1 | - | 10, 11 | 1660 | 6406821 | ||
D13801.1 | - | 5, 6 | 771 | |||
L13763.1 | - | 4, 5 | 605 |