Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fem1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010193.4 | NP_034323.1 | 2 | 844 | ||
GenBank mRNA | AB022863.1 | - | 2 | 754 | ||
AF064448.1 | - | 2 | 575 | |||
AF178633.1 | - | 2 | 501 | |||
AK032338.1 | - | 2 | 821 | |||
AK122272.1 | - | 1 | 42 | |||
AK132692.1 | - | 2 | 837 | |||
AK145371.1 | - | 1 | 207 | |||
AK149329.1 | - | 2 | 807 | |||
AK154060.1 | - | 2 | 1132 | |||
BC068236.1 | - | 2 | 798 | 30544277 |