Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fem1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010193.4 | NP_034323.1 | 2 | 2281 | ||
GenBank mRNA | AF064448.1 | - | 2 | 2012 | ||
AK032338.1 | - | 2 | 2258 | |||
AK078303.1 | - | 1 | 183 | |||
AK122272.1 | - | 1 | 1479 | |||
AK145371.1 | - | 1 | 1644 | |||
AK149329.1 | - | 2 | 2244 | |||
AK160393.1 | - | 1 | 217 | |||
BC068236.1 | - | 2 | 2235 | 30544277 |