Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010450.3 | NP_034580.1 | 2 | 1042 | ||
GenBank mRNA | AK133481.1 | - | 2 | 1042 | ||
BC052232.1 | - | 2 | 1019 | |||
BC119302.1 | - | 2 | 977 | 8734051 | ||
U20370.1 | - | 2 | 998 |