Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il18rap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010553.4 | NP_034683.1 | 7 | 1081 | ||
GenBank mRNA | AF077347.1 | - | 7 | 1090 | ||
AK137541.1 | - | 7 | 1042 | |||
BC120598.1 | - | 7 | 1088 | 40129521 | ||
BC120600.1 | - | 7 | 1088 | 40129523 |