Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdgfra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083316.1 | NP_001076785.1 | 3 | 548 | ||
NM_011058.2 | NP_035188.2 | 3 | 531 | |||
GenBank mRNA | AK035501.1 | - | 3 | 488 | ||
AK081664.1 | - | 3 | 530 | |||
AK136490.1 | - | 3 | 517 | |||
AK139094.1 | - | 2 | 763 | |||
AK145097.1 | - | 3 | 1131 | |||
AK147267.1 | - | 3 | 480 | |||
AK163952.1 | - | 3 | 595 | |||
BC020405.1 | - | 3 | 462 | |||
BC037467.1 | - | 3 | 470 | |||
BC053036.1 | - | 3 | 548 | 5704645 | ||
M57683.1 | - | 3 | 430 | |||
M84607.1 | - | 3 | 541 |