Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011318.2 | NP_035448.2 | 2 | 400 | ||
GenBank mRNA | AK144248.1 | - | 2 | 287 | ||
AK165699.1 | - | 2 | 287 | |||
BC061125.1 | - | 2 | 297 | 30308397 | ||
M23552.1 | - | 2 | 376 | |||
X14079.1 | - | 2 | 376 | |||
Y00426.1 | - | 2 | 389 |