Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011390.2 | NP_035520.1 | 2 | 218 | ||
GenBank mRNA | AF087436.1 | - | 2 | 220 | ||
AK052910.1 | - | 2 | 269 | |||
AK083598.1 | - | 2 | 176 | |||
AK131129.1 | - | 2 | 172 | |||
AK143535.1 | - | 2 | 212 | |||
AK149750.1 | - | 2 | 213 | |||
AK166215.1 | - | 2 | 212 | |||
AK169950.1 | - | 2 | 208 | |||
AK171498.1 | - | 2 | 255 | |||
BC051558.1 | - | 2 | 207 | |||
BC053470.1 | - | 2 | 844 | |||
BC059242.1 | - | 2 | 212 | |||
BC060133.1 | - | 2 | 212 | |||
BC079677.1 | - | 2 | 176 | |||
BC141107.1 | - | 2 | 178 | 9055719 |