Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011520.3 | NP_035650.2 | 3 | 517 | ||
GenBank mRNA | AK045898.1 | - | 3 | 303 | ||
AK129153.1 | - | 3 | 517 | |||
AK155127.1 | - | 3 | 502 | |||
BC054795.1 | - | 3 | 476 | 6834272 | ||
BC062093.1 | - | 3 | 413 | 5705893 | ||
U52826.1 | - | 3 | 417 |