Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011746.2 | NP_035876.2 | 1 | 402 | ||
GenBank mRNA | AK134870.1 | - | 1 | 417 | ||
BC054771.1 | - | 1 | 402 | 6415520 |