Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011755.2 | NP_035885.2 | 3 | 363 | ||
GenBank mRNA | AK132703.1 | - | 3 | 366 | ||
AK159080.1 | - | 3 | 317 | |||
BB614371.1 | - | 3 | 363 | |||
BC138830.1 | - | 3 | 282 | 8861852 | ||
BC138832.1 | - | 2 | 280 | 8861854 | ||
X17617.1 | - | 3 | 314 |