Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011773.3 | NP_035903.2 | 7 | 1095 | ||
GenBank mRNA | AK144568.1 | - | 7 | 1079 | ||
AK165796.1 | - | 7 | 1058 | |||
BC066199.1 | - | 7 | 1077 | 30476359 | ||
U76007.1 | - | 7 | 1053 |