Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011846.4 | NP_035976.3 | 3, 4 | 491 | ||
GenBank mRNA | AB021224.2 | - | 3, 4 | 497 | ||
AJ010731.1 | - | 3, 4 | 495 | |||
AK079221.1 | - | 262 | ||||
AK143521.1 | - | 3, 4 | 490 | |||
BC051917.1 | - | 3, 4 | 491 | 5716458 |