Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tenm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011855.4 | NP_035985.2 | 9 | 1549 | ||
GenBank mRNA | AB025410.1 | - | 9 | 1551 | ||
AK047423.1 | - | 11 | 2314 | |||
BC138860.1 | - | 8 | 1532 | 8861882 | ||
BC138861.1 | - | 8 | 1532 | 8861883 |